Learn More
Medchemexpress LLC FAM azide, 6-isomer | 1386385-76-7 | MFCD29912889 | 99.8% | 10 MG
SDP
Catalog No. 5000299050
Encompass_Preferred
FAM azide, 6-isomer is a fluorescein-derived azide labeling reagent used for fluorescent tagging and bioconjugation via click chemistry. It offers a bright fluorescein signal and an azide functional group for efficient conjugation to alkyne-containing molecules.
  • Fluorescent fluorescein-based dye with azide functional group.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO/BCN reagents.
  • Supplied as a solid; solution variants in DMSO are also available from some suppliers.
  • Molecular weight 458.42 and CAS 1386385-76-7; high purity for labeling applications.
  • Available in small pack sizes suitable for research-scale labeling.
Catalog No. 50-002-99050 Supplier Medchemexpress LLC Supplier No. HYD131410MG
FAM azide, 6-isomer is a fluorescein-derived azide labeling reagent used for fluorescent tagging and bioconjugation via click chemistry. It offers a bright fluorescein signal and an azide functional group for efficient conjugation to alkyne-containing molecules.
  • Fluorescent fluorescein-based dye with azide functional group.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO/BCN reagents.
  • Supplied as a solid; solution variants in DMSO are also available from some suppliers.
  • Molecular weight 458.42 and CAS 1386385-76-7; high purity for labeling applications.
  • Available in small pack sizes suitable for research-scale labeling.