Learn More
Cayman Chemical FAM Azide 6-isomer, Purity: ≥95%, CAS: 1386385-76-7, 5 mg
SDP
Catalog No. 502945237
Encompass_Preferred
A clickable form of 6-FAM; has been used to label EdU for use in cell viability, proliferation, and cell cycle progression assays in cancer cells; a related derivative, 6-FAM, displays excitation/emission maxima of 492/518 nm, respectively
Catalog No. 50-294-5237 Supplier Cayman Chemical Supplier No. 423895 MG
A clickable form of 6-FAM; has been used to label EdU for use in cell viability, proliferation, and cell cycle progression assays in cancer cells; a related derivative, 6-FAM, displays excitation/emission maxima of 492/518 nm, respectively