Learn More
CPC Scientific FAM-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific AMYD011A
Encompass
beta-Amyloid (1-40) found to exhibit both neurotrophic and neurotoxic effects that depend on neuronal age and beta-protein concentration.ONE-LETTER SEQUENCE: FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C215H305N53O64S1MOLECULAR WEIGHT:4688.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1678416-08-4], SYNONYMS: 5-FAM-Amyloid β-Protein (1-40)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: B.A. Yankner et al., Science, 250, 279 (1990)
Catalog No. 50-210-8659