Learn More
CPC Scientific 5FAM-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108679
Encompass
Synthetic peptide that forms fibrils with have the same antigenic and structural features as those of native AD amyloid filaments.ONE-LETTER SEQUENCE: 5FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKMOLECULAR FORMULA: C166H219N41O52MOLECULAR WEIGHT:3620.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [109770-29-8], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: J. Kang et al., Nature, 325, 773 (1987)
Catalog No. 50-210-8679 Supplier CPC Scientific Supplier No. AMYD025B
Synthetic peptide that forms fibrils with have the same antigenic and structural features as those of native AD amyloid filaments.ONE-LETTER SEQUENCE: 5FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKMOLECULAR FORMULA: C166H219N41O52MOLECULAR WEIGHT:3620.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [109770-29-8], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: J. Kang et al., Nature, 325, 773 (1987)