Learn More
CPC Scientific 5FAM-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108652
Encompass
42-residue peptide believed to be the major subunit of the vascular and plaque filaments in individuals with Alzheimer's disease, elderly people and patients with Trisomy 21 (Down's Syndrome). Inactive control.ONE-LETTER SEQUENCE: 5Fam-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIAMOLECULAR FORMULA: C224H321N55O66S1MOLECULAR WEIGHT:4872.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802087-78-0], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: Neurobiology of Aging, 13, No.5 (1992)
Catalog No. 50-210-8652 Supplier CPC Scientific Supplier No. AMYD005B
42-residue peptide believed to be the major subunit of the vascular and plaque filaments in individuals with Alzheimer's disease, elderly people and patients with Trisomy 21 (Down's Syndrome). Inactive control.ONE-LETTER SEQUENCE: 5Fam-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIAMOLECULAR FORMULA: C224H321N55O66S1MOLECULAR WEIGHT:4872.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802087-78-0], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: Neurobiology of Aging, 13, No.5 (1992)