Learn More
CPC Scientific 5FAM-Ser-Pro-Lys-Met-Val-Gln-Gly-Ser-Gly-Cys-Phe-Gly-Arg-Lys-Met-Asp-Arg-Ile-Ser-Ser-Ser-Ser-Gly-Leu-Gly-Cys-Lys-Val-Leu-Arg-Arg-His-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109858
Encompass
Brain natriuretic peptide or B-type natriuretic peptide (BNP) (also ventricular natriuretic peptide) is a 32-amino acid peptide secreted by heart ventricles in response to excessive stretching of heart muscle cells (cardiomyocytes).ONE-LETTER SEQUENCE: 5FAM-SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH (Cys10 and 26 bridge)MOLECULAR FORMULA: C164H256N50O48S4MOLECULAR WEIGHT:3824.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [114471-18-0], RESEARCH AREA: CardiovascularREFERENCES: T. Sudoh et al., BBRC, 159, 1427 (1989)
Catalog No. 50-210-9858 Supplier CPC Scientific Supplier No. NATR010A
Brain natriuretic peptide or B-type natriuretic peptide (BNP) (also ventricular natriuretic peptide) is a 32-amino acid peptide secreted by heart ventricles in response to excessive stretching of heart muscle cells (cardiomyocytes).ONE-LETTER SEQUENCE: 5FAM-SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH (Cys10 and 26 bridge)MOLECULAR FORMULA: C164H256N50O48S4MOLECULAR WEIGHT:3824.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [114471-18-0], RESEARCH AREA: CardiovascularREFERENCES: T. Sudoh et al., BBRC, 159, 1427 (1989)