Learn More
CPC Scientific 5FAM-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific BRAK002A
Encompass
ONE-LETTER SEQUENCE: 5FAM-RPPGFSPFRMOLECULAR FORMULA: C71H83N15O17MOLECULAR WEIGHT:1418.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1490132], RESEARCH AREA: CardiovascularREFERENCES: D. Regoli et al., J. Pharmacol. Rev., 32, 1 (1980)
Catalog No. 50-210-8847