Learn More
Medchemexpress LLC FAM-DEALA-Hyp-YIPD | 99.4% | 1477.48 | 5 MG
SDP
Catalog No. 5000396325
Encompass_Preferred
FAM-DEALA-Hyp-YIPD is a fluorescent HIF-1α peptide with a Kd of 180-560 nM.
  • Assesses VHL binding in Fluorescence Polarization (FP) displacement assays.
  • Evaluates the impact of VHL binding on degradation activity.
Catalog No. 50-003-96325 Supplier Medchemexpress LLC Supplier No. HYP5908F5MG
FAM-DEALA-Hyp-YIPD is a fluorescent HIF-1α peptide with a Kd of 180-560 nM.
  • Assesses VHL binding in Fluorescence Polarization (FP) displacement assays.
  • Evaluates the impact of VHL binding on degradation activity.