Learn More
Immunochemistry Technology FAM FLICA CASPASE 1 ASSAY KIT
Catalog No. NC0585813
Encompass
FAM-FLICA™ Caspase-1 Assay Kit, 25 Tests Detect caspase-1 activity with the FAM FLICA™ Caspase-1 Assay Kit. This in vitro assay employs the fluorescent inhibitor probe FAM-YVAD-FMK to label active caspase-1 enzyme in living cells or tissue samples. Analyze the green fluorescent signal using fluorescence microscopy, a fluorescent plate reader, or by flow cytometry. Caspases play important roles in apoptosis and inflammation. ICT's FLICA assay kits are used by researchers seeking to quantitate apoptosis via caspase activity in cultured cells and tissues. The FAM FLICA Caspase 1 probe allows researchers to assess caspase-1 activation.
Catalog No. NC0585813 Supplier Immunochemistry Technology Supplier No. 97
FAM-FLICA™ Caspase-1 Assay Kit, 25 Tests Detect caspase-1 activity with the FAM FLICA™ Caspase-1 Assay Kit. This in vitro assay employs the fluorescent inhibitor probe FAM-YVAD-FMK to label active caspase-1 enzyme in living cells or tissue samples. Analyze the green fluorescent signal using fluorescence microscopy, a fluorescent plate reader, or by flow cytometry. Caspases play important roles in apoptosis and inflammation. ICT's FLICA assay kits are used by researchers seeking to quantitate apoptosis via caspase activity in cultured cells and tissues. The FAM FLICA Caspase 1 probe allows researchers to assess caspase-1 activation.