Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FAM-labeled ODN 1826 sodium | 00-00-0 | 98.0% | 5 MG
SDP

Catalog No. 5000223025
Encompass_Preferred

FAM-labeled ODN 1826 sodium is a class B CpG oligodeoxynucleotide that functions as a TLR9 agonist and is labeled with fluorescein (FAM) for fluorescence-based studies. It is supplied as a light yellow solid for research use and is intended for evaluating cellular uptake and localization by confocal microscopy or flow cytometry.

  • Class B CpG oligodeoxynucleotide.
  • TLR9 agonist to stimulate innate immune responses.
  • Fluorescein (FAM) label enables fluorescent detection.
  • High purity (98.03%) and solid, light yellow appearance.
  • Storage: -20°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-23025 Supplier Medchemexpress LLC Supplier No. HY146245F5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FAM-labeled ODN 1826 sodium is a class B CpG oligodeoxynucleotide that functions as a TLR9 agonist and is labeled with fluorescein (FAM) for fluorescence-based studies. It is supplied as a light yellow solid for research use and is intended for evaluating cellular uptake and localization by confocal microscopy or flow cytometry.

  • Class B CpG oligodeoxynucleotide.
  • TLR9 agonist to stimulate innate immune responses.
  • Fluorescein (FAM) label enables fluorescent detection.
  • High purity (98.03%) and solid, light yellow appearance.
  • Storage: -20°C sealed; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.