Learn More
Lumiprobe FAM maleimide, 6-isomer, 25 mg
Catalog No. 501143190
Encompass_Preferred
FAM (fluorescein) is a bright fluorophore which is compatible with various fluorescence detection instruments. FAM is a universal dye that is useful for microscopy, qPCR and many other methods as well as for FRET-based and fluorescence polarization based binding assays.
Catalog No. 50-114-3190 Supplier Lumiprobe Supplier No. 44180
FAM (fluorescein) is a bright fluorophore which is compatible with various fluorescence detection instruments. FAM is a universal dye that is useful for microscopy, qPCR and many other methods as well as for FRET-based and fluorescence polarization based binding assays.