Learn More
Biotium FAM SE, 5 and 6 isomers
Catalog No. 501964805
Encompass_Preferred
5(6)-FAM SE (full name: 5-(and-6)-Carboxyfluorescein, succinimidyl ester mixed isomers) is an amine-reactive green fluorescent dye widely used for labeling proteins or other molecules that contain a primary or secondary aliphatic amine. The coupling reaction is usually carried out at pH 8-9.5. The amide linkage from the coupling reaction is much more stable than the thiourea linkage formed from the coupling of an amine and an isothiocyanate. Properties: Ex / Em = 495/519 nm; Yellow solid soluble in DMSO or DMF; Store at -20°C and protect from light; C 25 H 15 NO 9; MW: 473.4; [117548-22-8]
Catalog No. 50-196-4805 Supplier Biotium Supplier No. 90024
5(6)-FAM SE (full name: 5-(and-6)-Carboxyfluorescein, succinimidyl ester mixed isomers) is an amine-reactive green fluorescent dye widely used for labeling proteins or other molecules that contain a primary or secondary aliphatic amine. The coupling reaction is usually carried out at pH 8-9.5. The amide linkage from the coupling reaction is much more stable than the thiourea linkage formed from the coupling of an amine and an isothiocyanate. Properties: Ex / Em = 495/519 nm; Yellow solid soluble in DMSO or DMF; Store at -20°C and protect from light; C 25 H 15 NO 9; MW: 473.4; [117548-22-8]