Learn More
Aat Bioquest FAM-VAD-FMK (100 Tests)
SDP
Catalog No. 502249444
Encompass_Preferred
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1 2 3 6 8 9 or 10. This catalog is for 100 Tests.
This item is not returnable. View return policy
Catalog No. 50-224-9444 Supplier Aat Bioquest Supplier No. 13484
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1 2 3 6 8 9 or 10. This catalog is for 100 Tests.