Learn More
Aviva Systems Biology Corp RFLNA Antibody - middle region (ARP53404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM101A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM101A. Alternate names: FLJ44614ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-61018