Learn More
Aviva Systems Biology Corp FAM107A Antibody - middle region (ARP42348_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42348_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM107A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM107A. Alternate names: DRR1, FLJ30158, FLJ45473, TU3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58192