Learn More
Aviva Systems Biology Corp Fam114a2 Antibody - C-terminal region (ARP52384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fam114a2. It was validated on Western Blot. . Alternate names: 1810073G14Rik, 9030624B09Rik, RP23-206L21.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-711