Learn More
Aviva Systems Biology Corp FAM118A Antibody - middle region (ARP56314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM118A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FAM118A. Alternate names: C22orf8, FLJ20635ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-138