Learn More
Aviva Systems Biology Corp FAM119A Antibody - N-terminal region (ARP52843_P050) | 100uL
SDP
Catalog No. 5001460847
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM119A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM119A. Alternate names: MGC45373, FAM119A, HCA557bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60847 Supplier Aviva Systems Biology Corp Supplier No. ARP52843_P050
This is a rabbit polyclonal antibody against FAM119A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM119A. Alternate names: MGC45373, FAM119A, HCA557bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat