Learn More
Aviva Systems Biology Corp FAM119A Antibody - middle region (ARP52844_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM119A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM119A. Alternate names: MGC45373, FAM119A, HCA557bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60848