Learn More
Aviva Systems Biology Corp FAM120A Antibody - middle region (ARP55089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM120A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM120A. Alternate names: C9orf10, DNAPTP1, DNAPTP5, MGC111527, MGC133257, OSSAApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53747