Learn More
Aviva Systems Biology Corp Fam122b Antibody - N-terminal region (ARP52850_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52850_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fam122b. It was validated on Western Blot. . Alternate names: 4632404H22Rik, AI256344, SPACIA2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-775