Learn More
Aviva Systems Biology Corp Fam123a Antibody - N-terminal region (ARP55491_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55491_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fam123a. It was validated on Western Blot. . Alternate names: 2600011E07Rik, Amer2, Fam123aApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-286-044