Learn More
Aviva Systems Biology Corp FAM124A Antibody - middle region (ARP55453_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55453_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM124A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM124A. Alternate names: FLJ30707ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61510