Learn More
Aviva Systems Biology Corp FAM129A Antibody - N-terminal region (ARP42692_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42692_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM129A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM129A. Alternate names: C1orf24, FLJ38228, NIBAN, GIG39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58309