Learn More
Aviva Systems Biology Corp FAM133B Antibody - C-terminal region (ARP54509_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54509_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM133B. It was validated on Western Blot. . Alternate names: MGC40405ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-934