Learn More
Aviva Systems Biology Corp RETREG1 Antibody - middle region (ARP44827_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44827_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM134B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human FAM134B. Alternate names: FLJ20152, FLJ22155, FLJ22179, JK1, JK-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53436