Learn More
Aviva Systems Biology Corp RETREG1 Antibody - middle region (ARP44826_P050) | 100uL
SDP
Catalog No. 5001458907
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM134B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM134B. Alternate names: FLJ20152, FLJ22155, FLJ22179, JK1, JK-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58907 Supplier Aviva Systems Biology Corp Supplier No. ARP44826_P050
This is a rabbit polyclonal antibody against FAM134B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM134B. Alternate names: FLJ20152, FLJ22155, FLJ22179, JK1, JK-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast