Learn More
Aviva Systems Biology Corp FAM135B Antibody - middle region (ARP56771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM135B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM135B. Alternate names: C8ORFK32, MGC126009, MGC126010, MGC33221ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61984