Learn More
Aviva Systems Biology Corp FAM13C1 Antibody - N-terminal region (ARP54376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM13C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM13C1. Alternate names: MGC33233, FAM13C1ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61192