Learn More
Aviva Systems Biology Corp FAM14A Antibody - middle region (ARP50028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM14A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM14A. Alternate names: MGC44913, TLH29, FAM14A, ISG12BApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-60246