Learn More
Aviva Systems Biology Corp FAM156A Antibody - N-terminal region (ARP40755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM156A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM156A. Alternate names: DKFZp686B22211, PRO0659, TMEM29, FAM156BApplicationWB. Species reactivity: Human
Catalog No. 50-014-57612