Learn More
Aviva Systems Biology Corp ABRAXAS1 Antibody - C-terminal region (ARP65319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM175A. It was validated on Western Blot. . Alternate names: ABRA1, CCDC98, FLJ11520, FLJ12642, FLJ13614ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-288-472