Learn More
Aviva Systems Biology Corp Fam176b Antibody - middle region (ARP42509_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42509_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fam176b. It was validated on Western Blot. . Alternate names: RGD1308876, Fam176bApplicationWB. Species reactivity: Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-006