Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inosine 5'-Monophosphate Disodium Salt, Hydrate, Spectrum™ Chemical
SDP

Catalog No. 18620948 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-948 10 kg
18-620-945 5 g
18-620-946 25 g
18-620-947 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-948 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I304110KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: 5'-Inosinic Acid Disodium Salt Hydrate, Disodium 5'-Inosinate Monohydrate, 5'-Inosinic Acid Disodium Hydrate

5'-Inosinic Acid Disodium Salt Hydrate, Disodium 5'-Inosinate Monohydrate, 5'-Inosinic Acid Disodium Hydrate

Chemical Identifiers

CAS 4691-65-0

Specifications

Packaging Drum
Quantity 10 kg
Molecular Weight (g/mol) 392.17
Percent Purity 98%
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Inosine 5'-Monophosphate Disodium Salt, Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.