Learn More
Aviva Systems Biology Corp FAM207A Antibody - N-terminal region (ARP45896_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45896_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM207A. It was validated on Western Blot. . Alternate names: PRED56, C21orf70ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig
Catalog No. 50-285-228