Learn More
Aviva Systems Biology Corp FAM29A Antibody - middle region (ARP56990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM29A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM29A. Alternate names: MGC102696, MGC138798, MGC138799, RP11-296P7.3, Dgt6, FAM29AApplicationWB. Species reactivity: Horse, Human, Rabbit
Catalog No. 50-014-62055