Learn More
Aviva Systems Biology Corp FAM29A Antibody - middle region (ARP56989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM29A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FAM29A. Alternate names: Dgt6, FAM29A, MGC102696, MGC138798, MGC138799, RP11-296P7.3ApplicationWB. Species reactivity: Human
Catalog No. 50-286-251