Learn More
Aviva Systems Biology Corp SHLD2 Antibody - N-terminal region (ARP57321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM35A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM35A. Alternate names: MGC5560, bA163M19.1, FAM35A1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53950