Learn More
Aviva Systems Biology Corp FAM38B Antibody - middle region (ARP49683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM38B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM38B. Alternate names: FLJ23144, FLJ23403, HsT748, PIEZO2, C18orf30, C18orf58, FAM38B, HsT771, FAM38B2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53596