Learn More
Aviva Systems Biology Corp FAM3D Antibody - middle region (ARP52652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM3D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM3D. Alternate names: EF7, OIT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-748