Learn More
Medchemexpress LLC FAM49B (190-198) mouse | 1588388-60-6 | 1042.20 | 10 MG
SDP
Catalog No. 5000424916
Encompass_Preferred
FAM49B (190-198) mouse is a peptide fragment of FAM49B. FAM49B is a mitochondria-localized protein that regulates mitochondrial fission, function and integrity, and tumor progression. It also acts as a negative regulator in T cell activation by repressing GTPase Rac activity and modulating cytoskeleton reorganization.
  • Regulates mitochondrial fission
  • Involved in mitochondrial function and integrity
  • Contributes to tumor progression
  • Acts as a negative regulator in T cell activation
  • Represses GTPase Rac activity
  • Modulates cytoskeleton reorganization
  • Soluble in DMSO at 50 mg/mL
  • Store as powder at -80°C for 2 years or -20°C for 1 year in sealed, moisture-free conditions
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed, moisture-free conditions
Catalog No. 50-004-24916 Supplier Medchemexpress LLC Supplier No. HYP1065010MG
Add to Cart
Add to Cart
FAM49B (190-198) mouse is a peptide fragment of FAM49B. FAM49B is a mitochondria-localized protein that regulates mitochondrial fission, function and integrity, and tumor progression. It also acts as a negative regulator in T cell activation by repressing GTPase Rac activity and modulating cytoskeleton reorganization.
  • Regulates mitochondrial fission
  • Involved in mitochondrial function and integrity
  • Contributes to tumor progression
  • Acts as a negative regulator in T cell activation
  • Represses GTPase Rac activity
  • Modulates cytoskeleton reorganization
  • Soluble in DMSO at 50 mg/mL
  • Store as powder at -80°C for 2 years or -20°C for 1 year in sealed, moisture-free conditions
  • Store in solvent at -80°C for 6 months or -20°C for 1 month in sealed, moisture-free conditions