Learn More
Aviva Systems Biology Corp FAM50B Antibody - N-terminal region (ARP54850_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54850_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM50B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM50B. Alternate names: D6S2654E, X5LApplicationWB. Species reactivity: Human
Catalog No. 50-014-61343