Learn More
Aviva Systems Biology Corp FAM53A Antibody - C-terminal region (ARP51825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM53A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FAM53A. Alternate names: DNTNPApplicationWB. Species reactivity: Human
Catalog No. 50-285-598