Learn More
Aviva Systems Biology Corp FAM54A Antibody - C-terminal region (ARP51900_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51900_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM54A. It was validated on Western Blot. . Alternate names: DUFD1, FAM54AApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-285-606