Learn More
Aviva Systems Biology Corp FAM54A Antibody - middle region (ARP51899_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51899_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM54A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM54A. Alternate names: DUFD1, FAM54AApplicationWB. Species reactivity: Cow, Human, Rabbit
Catalog No. 50-014-53678