Learn More
Aviva Systems Biology Corp Fam55d Antibody - C-terminal region (ARP45014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fam55d. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: C130036J11, D930028F11Rik, Fam55dApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-176