Learn More
Aviva Systems Biology Corp MINDY1 Antibody - middle region (ARP56272_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56272_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM63A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM63A. Alternate names: FLJ11280, FLJ43504, RP11-316M1.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61789