Learn More
Aviva Systems Biology Corp FAM70A Antibody - N-terminal region (ARP47380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM70A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM70A. Alternate names: FLJ20716, RP3-525N14.6, FAM70AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59530