Learn More
Aviva Systems Biology Corp FAM71A Antibody - C-terminal region (ARP53187_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53187_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM71A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FAM71A. Alternate names: FLJ32796, RP11-338C15.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rabbit
Catalog No. 50-285-805