Learn More
Aviva Systems Biology Corp FAM71B Antibody - middle region (ARP52552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM71B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM71B. Alternate names: MGC26988ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-60771